Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Galactosidase Rabbit anti-E. coli, Alexa Fluor™ 350, Clone: 13R4, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP252652AF350

Catalog No. NP2262A30


Add to cart

Description

Description

beta Galactosidase Monoclonal antibody specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot,Flow Cytometry.
Specifications

Specifications

beta Galactosidase
13R4
Western Blot, Flow Cytometry
EC 3.2.1.23; b-gal
Beta-galactosidase.
0.1 mL
Primary
13R4 recognizes wild-type E. coli beta-galactosidase.
Flow Cytometry, Western Blot
Alexa Fluor™ 350
50 mM sodium borate with 0.05% sodium azide
Rabbit
IgG Lambda
Store at 4°C in the dark.
Monoclonal
E. coli
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit