Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
beta Galactosidase Antibody (13R4), Alexa Fluor™ 532 - Chimeric, Novus Biologicals™
Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP252652AF532
Description
beta Galactosidase Monoclonal specifically detects beta Galactosidase in E. coli samples. It is validated for Western Blot, Flow Cytometry.Specifications
beta Galactosidase | |
Monoclonal | |
Alexa Fluor 532 | |
50 mM sodium borate with 0.05% sodium azide | |
Rabbit | |
Protein A or G purified | |
Primary | |
E. coli | |
IgG λ |
Western Blot, Flow Cytometry | |
13R4 | |
Western Blot, Flow Cytometry | |
EC 3.2.1.23; b-gal | |
Beta-galactosidase. | |
0.1 mL | |
13R4 recognizes wild-type E. coli beta-galactosidase. | |
Store at 4°C in the dark. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction