Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Beta Ig-h3/TGFBI Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications
$416.50 - $697.00
Specifications
Antigen | Beta Ig-h3/TGFBI |
---|---|
Dilution | Simple Western 1:25, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 |
Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
Beta Ig-h3/TGFBI Polyclonal specifically detects Beta Ig-h3/TGFBI in Human samples. It is validated for Simple Western, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
Beta Ig-h3/TGFBI | |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
Beta ig-h3, BIGH3EBMD, CDB1, CDG2, CDGG1transforming growth factor, beta-induced, 68kD, CSD, CSD1, CSD2, CSD3, Kerato-epithelin, LCD1, RGD-CAP, RGD-containing collagen-associated protein, transforming growth factor, beta-induced, 68kDa, transforming growth factor-beta-induced protein ig-h3 | |
TGFBI | |
IgG | |
Affinity Purified | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Simple Western 1:25, Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
Polyclonal | |
Rabbit | |
Angiogenesis, Cytokine Research, Immunology, Vision | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
7045 | |
This antibody was developed against Recombinant Protein corresponding to amino acids:DHHATNGVVHLIDKVISTITNNIQQIIEIEDTFETLRAAVAASGLNTMLEGNGQYTLLAPTNEAFEKIPSETLNRILGDPEALRDLLNNHILKSAMCAEAIVAGLSVETLEGTTLEVGCSGDMLTINGKAIISNKD | |
Primary | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title