Learn More
beta II Tubulin A Mouse anti-Human, DyLight 350, Clone: 6A10B2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237376UV
Catalog No. NBP237376UV
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
beta II Tubulin A
6A10B2
Western Blot, Flow Cytometry, ELISA
Purified
TUBB2A
Purified recombinant fragment of human TUBB2A (AA: 25-187) expressed in E. Coli.
Protein A or G purified
Apoptosis, Cell Biology, Cytoskeleton Markers
Primary
7280
ELISA, Flow Cytometry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
dJ40E16.7, TUBB, TUBB2, TUBB2B, tubulin beta-2A chain, tubulin, beta 2, tubulin, beta 2A, tubulin, beta polypeptide, tubulin, beta polypeptide 2
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human