Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta II Tubulin A Mouse anti-Human, HRP, Clone: 6A10B2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237376H

Catalog No. NBP237376H

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

beta II Tubulin A
6A10B2
Western Blot, ELISA
Purified
TUBB2A
Purified recombinant fragment of human TUBB2A (AA: 25-187) expressed in E. Coli.
Protein A or G purified
Apoptosis, Cell Biology, Cytoskeleton Markers
Primary
7280
ELISA, Western Blot
HRP
PBS with No Preservative
dJ40E16.7, TUBB, TUBB2, TUBB2B, tubulin beta-2A chain, tubulin, beta 2, tubulin, beta 2A, tubulin, beta polypeptide, tubulin, beta polypeptide 2
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit