Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Lactose, Anhydrous, 99%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
Packaging:
Amber Glass Bottle
5965-66-2
C12H22O11
342.30
GUBGYTABKSRVRQ-DCSYEGIMSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
5965-66-2
C12H22O11
342.30
GUBGYTABKSRVRQ-DCSYEGIMSA-N
(2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-{[(2R,3S,4R,5R,6R)-4,5,6-trihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy}oxane-3,4,5-triol
OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O

L3147, 5965-66-2, C12H22O11

Spectrum™ beta-Lactose, Anhydrous , also known as milk sugar or 4-O-Β-D-galactopyranosyl-D-glucose, is a disaccharide sugar and is added to pills as a filler because of its physical properties. Anhydrous products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 500 g
CAS Max % 1
Percent Purity 99%