Alfa Aesar™ beta-Naphthol Violet

Manufacturer: Alfa Aesar™ B2427918

Quantity 50g

 View more versions of this product

Catalog No. AAB2427918

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7143-21-7
    Molecular Formula C16H9N3Na2O9S2
    Formula Weight 497.37
    MDL Number MFCD00021457
    Chemical Name or Material beta-Naphthol Violet

    Specifications

    Formula Weight 497.37
    Quantity 50g
    Molecular Formula C16H9N3Na2O9S2
    MDL Number MFCD00021457
    CAS 7143-21-7
    Health Hazard 1 Warning
    Odor Odorless
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Chemical Name or Material beta-Naphthol Violet

    Description

    Applications
    Beta-Naphthol violet exhibits antiseptic properties and is used in the synthesis of certain azo dyes and antioxidants for rubber. It can also be used in the detection of the ferric ion which turns the Beta-Naphthol violet solution green.

    Solubility
    Soluble in water.

    Notes
    Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit