Learn More
beta-NGF Mouse anti-Bovine, Clone: AS18, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP266610
Catalog No. NBP266610
Add to cart
Description
Description
beta-NGF Monoclonal antibody specifically detects beta-NGF in Bovine samples. It is validated for Immunoprecipitation, Enzyme Immunoassay, Neutralization.
Specifications
Specifications
beta-NGF
AS18
Immunoprecipitation 1:10 - 1:500, Enzyme Immunoassay, Neutralization
Purified
NGF
Hybridoma produced by the fusion of splenocytes from BALB/c mice immunized with human recombinant nerve growth factor and mouse myeloma cells.
Protein A or G purified
Cancer, Neuroscience
Primary
4803
ELISA, Immunoprecipitation, Neutralization
Unconjugated
PBS with No Preservative
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Mouse
IgG1
500μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Bovine, Human, Mouse, Reptile