Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-NGF Mouse anti-Bovine, Clone: AS21, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP266609

Catalog No. NBP266609


Add to cart

Description

Description

beta-NGF Monoclonal antibody specifically detects beta-NGF in Bovine samples. It is validated for Enzyme Immunoassay, Neutralization.
Specifications

Specifications

beta-NGF
AS21
Enzyme Immunoassay, Neutralization
Purified
NGF
Human recombinant nerve growth factor (hNGF)
Protein A or G purified
Cancer, Neuroscience
Primary
4803
ELISA, Neutralization
Unconjugated
PBS with 0.08% Sodium Azide
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Mouse
IgG1
500μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Bovine, Human, Mouse, Reptile
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit