Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-NGF Rabbit anti-Human, Alexa Fluor 647, Clone: B64, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP289766AF647

Catalog No. NB067878


Add to cart

Description

Description

beta-NGF Monoclonal antibody specifically detects beta-NGF in Human samples. It is validated for ELISA
Specifications

Specifications

beta-NGF
B64
ELISA
Purified
Rabbit
IgG
0.1 mL
Cancer, Neuroscience
Primary
4803
ELISA
Alexa Fluor 647
50mM Sodium Borate
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Recombinant Human beta-NGF Protein
Protein A purified
RUO
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit