Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-NGF Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30354720UL

 View more versions of this product

Catalog No. NB094773


Add to cart

Description

Description

beta-NGF Polyclonal antibody specifically detects beta-NGF in Human, Mouse samples. It is validated for Western Blot, KnockDown
Specifications

Specifications

beta-NGF
Unconjugated
Purified
Rabbit
IgG
20 μL
Cancer, Neuroscience
Primary
4803
KnockDown, Western Blot
PBS with 50% glycerol, pH7.3.
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Recombinant fusion protein containing a sequence corresponding to amino acids 19-241 of human beta-NGF (NP_002497.2).
Affinity purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit