Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-NGF Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen beta-NGF
Applications KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB094772
View Documents
Novus Biologicals
NBP303547100UL
100 μg
Each for $320.71
Add to cart
 
NB094773
View Documents
Novus Biologicals
NBP30354720UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

beta-NGF Polyclonal antibody specifically detects beta-NGF in Human, Mouse samples. It is validated for Western Blot, KnockDown
Specifications

Specifications

beta-NGF
Unconjugated
Rabbit
Cancer, Neuroscience
PBS with 50% glycerol, pH7.3.
4803
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
KnockDown, Western Blot
Purified
RUO
Human, Mouse
Beta nerve growth factor precursor, beta-nerve growth factor, Beta-NGF, HSAN5, MGC161426, nerve growth factor (beta polypeptide), nerve growth factor, beta subunit, NGFBMGC161428
Recombinant fusion protein containing a sequence corresponding to amino acids 19-241 of human beta-NGF (NP_002497.2).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit