Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Nicotinamide Adenine Dinucleotide, Affymetrix/USB™

Click to view available options
Quantity:
1g
5g
10g
25g
100g
Packaging:
Plastic bottle
53-84-9
C21H26N7O14P2-
662.422
MFCD00036253
BAWFJGJZGIEFAR-NNYOXOHSSA-M
nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad +
15938971
CHEBI:57540
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O)C(=O)N
53-84-9
C21H26N7O14P2-
662.422
MFCD00036253
BAWFJGJZGIEFAR-NNYOXOHSSA-M
nicotinamide adenine dinucleotide,nad+,diphosphopyridine nucleotide,nad-oxidized,nicotinamide-adenine dinucleotide,dpn-ox,beta-nicotinamide adenine dinucleotide,dpn+,nad,nad +
15938971
CHEBI:57540
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-oxidophosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)([O-])OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O)C(=O)N
Packaging Plastic bottle
Quantity 5g
Formula Weight 717.47
Physical Form Powder
Chemical Name or Material beta-Nicotinamide Adenine Dinucleotide