Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-Nicotinamide adenine dinucleotide monosodium salt dihydrate, Affymetrix/USB™

Click to view available options
Quantity:
5g
100g
200g
Packaging:
Plastic bottle
20111-18-6
C21H27N7NaO14P2
686.419
MFCD00148917
DNHZOFBMPSMMFX-QYZPTAICSA-N
adenosine5'-trihydrogen diphosphate , p'&
131875671
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O)C(=O)N.[Na]
20111-18-6
C21H27N7NaO14P2
686.419
MFCD00148917
DNHZOFBMPSMMFX-QYZPTAICSA-N
adenosine5'-trihydrogen diphosphate , p'&
131875671
[[(2R,3S,4R,5R)-5-(6-aminopurin-9-yl)-3,4-dihydroxyoxolan-2-yl]methoxy-hydroxyphosphoryl] [(2R,3S,4R,5R)-5-(3-carbamoylpyridin-1-ium-1-yl)-3,4-dihydroxyoxolan-2-yl]methyl phosphate;sodium
C1=CC(=C[N+](=C1)C2C(C(C(O2)COP(=O)([O-])OP(=O)(O)OCC3C(C(C(O3)N4C=NC5=C4N=CN=C5N)O)O)O)O)C(=O)N.[Na]
Formula Weight 721.44
Physical Form Powder
Packaging Plastic bottle
Quantity 5g
Chemical Name or Material beta-Nicotinamide Adenine Dinucleotide Monosodium Salt Dihydrate