Learn More
beta-Nicotinamide adenine dinucleotide phosphate monosodium salt, Thermo Scientific Chemicals
Description
β-Nicotinamide adenine dinucleotide phosphate monosodium salt acts as an electron acceptor used by cells. It is also useful in vitro to study NADP. Furthermore, it is used in a wide range of enzyme catalyzed oxidation reduction reactions. The combination of NADP+/NADPH is employed in a variety of antioxidation mechanisms, which protect against reactive oxidation species accumulation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Color | White |
Quantity | 1 g |
Beilstein | 4779954 |
Solubility Information | Soluble in water. |
Formula Weight | 765.4 |
Physical Form | Powder |
Chemical Name or Material | beta-Nicotinamide adenine dinucleotide phosphate monosodium salt |
Safety and Handling
Hazard Category | Warning |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | GHS P Statement P261-P280-P305+P351+P338-P304+P340-P405-P501a Avoid breathing dust/fume/gas/mist/vapors/spray. Wear protective gloves/protective clothing/eye protection/face protection. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations. |
EINECSNumber | 214-664-6 |
TSCA | No |
Recommended Storage | Store at -20°C |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.