Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beta-Nicotinamide Adenine Dinucleotide, Reduced Dipotassium Salt, Approx. 95%, Spectrum™

Click to view available options
Quantity:
500 mg
1 g
Packaging:
Amber Glass Bottle
104809-32-7
104809-32-7

N2007, 104809-32-7, C21H27K2N7O14P2

Spectrum™ Beta-Nicotinamide Adenine Dinucleotide, Reduced Dipotassium Salt , is a coenzyme and a dinucleotide involved in redox reactions and can be used an oxidizing agent. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.
Packaging Amber Glass Bottle
Quantity 500 mg
CAS Max % 100%
Percent Purity Approx. 95%