Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta-TrCP1/BTRC Mouse anti-Human, Alexa Fluor™ 647, Clone: OTI3D5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP271401AF647

Catalog No. N271401A647


Add to cart

Description

Description

beta-TrCP1/BTRC Monoclonal antibody specifically detects beta-TrCP1/BTRC in Human samples. It is validated for Western Blot, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

beta-TrCP1/BTRC
OTI3D5
Western Blot, Immunocytochemistry/Immunofluorescence
Affinity Purified, Conjugated
BTRC
Recombinant protein fragment corresponding to amino acids 52-354 of human beta-TrCP1/BTRC (NP_378663) produced in E.coli.
Immunogen affinity purified
Signal Transduction, Vision
Primary
8945.0
Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor 647
50mM Sodium Borate with 0.05% Sodium Azide
beta-transducin repeat containing, betaTrCP, BETA-TRCP, beta-TrCP1, bTrCP, bTrCP1, E3RSIkappaB, F-box and WD repeats protein beta-TrCP, F-box/WD repeat-containing protein 1A, FBW1A, FBXW1, FBXW1ABTRCP, Fwd1, MGC4643, pIkappaBalpha-E3 receptor subunit
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit