Learn More
Aat BioquestSupplier Diversity Partner Beta-trifluoromethylumbelliferone [7-Hydroxy-4-trifluoromethylcoumarin] *CAS 575-03-1* DFS Item
Manufacturer:  Aat BioquestSupplier Diversity Partner 82
This coumarin compound is used as a calibration standard for 7-hydroxy-4-trifluoromethylcoumarin-based enzyme substrates. It has pH-dependent fluorescence.
Catalog No. 50-195-2701