Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.
beta Tubulin Mouse anti-Human, Mouse, Rat, Avian, Chicken, Primate, Rabbit, Alexa Fluor™ 750, Clone: BT7R, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP237830AF750
Catalog No. NP237830A70
Description
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human,Mouse,Rat,Avian,Chicken,Primate,Rabbit samples. It is validated for Western Blot,Flow Cytometry,ELISA,Immunohistochemistry,Immunocytochemistry,Immunofluorescence,Immunohistochemistry (Paraffin).
Specifications
Specifications
beta Tubulin
BT7R
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Flow (Intracellular)
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Mouse
IgG2a
Store at 4°C in the dark.
Monoclonal
Detects beta-tubulin in human, monkey, mouse, rat, rabbit and chicken samples with ELISA, Western blot and immunofluorescence applications.
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor™ 750
50 mM sodium borate with 0.05% sodium azide
TUBB
KLH conjugated peptide of the N-terminus of Beta-Tubulin.
0.1 mL
Primary
203068
Avian, Chicken, Human, Mouse, Primate, Rabbit, Rat