Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Tubulin, Rabbit anti-Human, Clone: S11B, Novus Biologicals™

Rabbit Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP2810640.025MG

 View more versions of this product

Catalog No. NB015203


Add to cart

Description

Description

beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

beta Tubulin
S11B
Unconjugated
PBS
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Rabbit
IgG Lambda
0.025 mg
Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary
203068
Human
Western Blot, ELISA
1 mg/mL
Western Blot, ELISA
Purified
TUBB
Human MBP (microtubule-binding protein).
Protein A purified
RUO
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Monoclonal
Binds specifically to human beta Tubulin (no crossreactivity with alpha-tubulin).
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit