Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

beta Tubulin, Rabbit anti-Human, Clone: S11B, Novus Biologicals™

Rabbit Monoclonal Antibody

$156.50 - $419.00

Specifications

Antigen beta Tubulin
Clone S11B
Applications Western Blot, ELISA
Host Species Rabbit
Target Species Human
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB015203
View Documents
Novus Biologicals
NBP2810640.025MG
0.025 mg
Each for $156.50
Add to cart
 
NB015204
View Documents
Novus Biologicals
NBP2810640.2MG
0.2 mg
Each for $419.00
Add to cart
 
Description

Description

beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human Samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

beta Tubulin
Western Blot, ELISA
Human
Primary
S11B
Rabbit
Monoclonal
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit