Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 18 HPLC Columns, 5µm Particle Size

Available in limited dimensions. Please contact Customer Service for more details.

Manufacturer:  Thermo Scientific™ 71505152130

 View more versions of this product

Catalog No. 71505152130


Add to cart

Description

Description

Thermo ScientificTM BetaBasicTM 18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Ordering Information:

Other column dimensions are also available. Please call Customer Service for more information.

Specifications

Specifications

Reversed Phase
Reversed Phase
13%
2.1 mm
150 mm
2 to 12
200 m²/g
C18
Yes
Silica
Analytical Column
LC/MS, APCI
5 µm
150 Å
L1
Beta
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit