Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ BetaBasic™ 8 HPLC Columns, 5µm Particle Size

Thermo ScientificTM BetaBasicTM 8 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

$852.00

Specifications

Carbon Load 7%
For Use With LC/MS, APCI
Stationary Phase C8
Surface Area 200 m²/g
USP Type L7
View More Specs

Products 1
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Particle Size Diameter (Metric) Length (Metric) Price Quantity & Availability  
71405154630
View Documents
Thermo Scientific™
71405154630
5 µm 4.6 mm 150 mm Each for $852.00
Add to cart
 
Description

Description

Thermo ScientificTM BetaBasicTM 8 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

  • Pore size suitable for small molecules, peptides and protein digests
  • Superb pH stability

Ordering Information:

Other column dimensions are also available. Please call Customer Service for more information.

Specifications

Specifications

7%
C8
L7
Yes
150 Å
LC/MS, APCI
200 m²/g
Reversed Phase
5 µm
2 to 9
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit