Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaine hydrochloride, 99%, extra pure, specified according to the requirements of U, ACROS Organics™

Click to view available options
Quantity:
250g
1kg
2.5kg
Packaging:
Plastic bottle
590-46-5
C5H12ClNO2
153.606
MFCD00011903
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
betaine hydrochloride,betaine chloride,pluchine,acidogeno,acipepsol,achylin,acidin,acinorm,acidol,acidine
11545
carboxymethyl(trimethyl)azanium;chloride
C[N+](C)(C)CC(=O)O.[Cl-]
590-46-5
C5H12ClNO2
153.606
MFCD00011903
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
betaine hydrochloride,betaine chloride,pluchine,acidogeno,acipepsol,achylin,acidin,acinorm,acidol,acidine
11545
carboxymethyl(trimethyl)azanium;chloride
C[N+](C)(C)CC(=O)O.[Cl-]
pH 0.8 to 1.2 (1:4)
Color Colorless to White
Infrared Spectrum Authentic
Quantity 2.5kg
Ignition Residue 0.1% max.
Water 0.5% max.
Identification A solution (1 in 20) responds to the test for chloride: Complies
Packaging Plastic bottle
Melting Point 241.0°C
CAS Min % 98.5
CAS Max % 100.0
Assay Percent Range 99%
Linear Formula (CH3)3NCH2CO2H·Cl
Merck Index 15, 1181
Formula Weight 153.61
Physical Form Powder or Crystals
Percent Purity 99%
Grade Extra Pure
Chemical Name or Material βine hydrochloride
Show More Show Less