Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaine Hydrochloride, USP, 98-100.5%, Spectrum™

Click to view available options
Quantity:
100 g
500 g
1 kg
12 kg
50 kg
Packaging:
Drum
Poly Bottle
Poly Pail
590-46-5
C5H12ClNO2
153.61
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
(carboxymethyl)trimethylazanium chloride
[Cl-].C[N+](C)(C)CC(O)=O
590-46-5
C5H12ClNO2
153.61
HOPSCVCBEOCPJZ-UHFFFAOYSA-N
(carboxymethyl)trimethylazanium chloride
[Cl-].C[N+](C)(C)CC(O)=O

B1104, 590-46-5, C5H11NO2•HCl

Spectrum™ Betaine Hydrochloride, USP is a vitamin-like substance used to treat hypochlorhydria. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.1%
Packaging Poly Bottle
Quantity 100 g
CAS Max % 100%
Percent Purity 98 to 100.5%