Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betamethasone, Powder, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
100 mg
500 mg
1 g
5 g
25 g
100 g
Packaging:
Amber Glass Bottle
378-44-9
C22H29FO5
392.47
UREBDLICKHMUKA-DVTGEIKXSA-N
(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO
378-44-9
C22H29FO5
392.47
UREBDLICKHMUKA-DVTGEIKXSA-N
(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-1,10-dihydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-7-one
C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]2(C)[C@@]1(O)C(=O)CO

BE180, 378-44-9, C22H29FO5

Spectrum™ Betamethasone, Powder, USP is a corticosteroid used to relieve skin irritation. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Loss on Drying 1%
Ignition Residue 0.2%
Packaging Amber Glass Bottle
Specific Rotation +118° to +126°
Quantity 100 g
CAS Max % 100%
Percent Purity 97 to 103%