Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betamethasone Valerate, Micronized Powder, USP, 97-103%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
2152-44-5
C27H37FO6
476.59
SNHRLVCMMWUAJD-SUYDQAKGSA-N
(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl pentanoate
CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
2152-44-5
C27H37FO6
476.59
SNHRLVCMMWUAJD-SUYDQAKGSA-N
(1R,2S,3aS,3bS,9aS,9bR,10S,11aS)-9b-fluoro-10-hydroxy-1-(2-hydroxyacetyl)-2,9a,11a-trimethyl-7-oxo-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl pentanoate
CCCCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO

BE185, 2152-44-5, C27H37FO6

Spectrum™ Betamethasone Valerate, Micronized Powder, USP is a glucocorticoid steroid applied as a topical cream, ointment, lotion or gel to treat itching and other minor skin conditions. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
CAS Max % 1
Percent Purity 97 to 103%
Packaging Amber Glass Bottle
Quantity 1 kg