Learn More
Thermo Scientific™ BetaSil™ Silica Guard Cartridges
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
Manufacturer:  Thermo Scientific™ 70005012101
 View more versions of this product
Catalog No. 70005012101
Add to cart
Description
Description
Experience a powerful column for separating non-polar and moderately polar compounds based on 100Å high purity silica and designed for small molecule analysis.
  • 100Å silica
  • High surface area
Specifications
Specifications
2 to 9
10 mm
2.1 mm
HPLC Column
L3
5 μm
100Å
2.1 mm
No
BetaSil
SDS
Documents
Product Certifications
Accessories