Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Betaxolol hydrochloride, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
10mg
50mg

Chemical Identifiers

63659-19-8
C18H30ClNO3
343.892
CHDPSNLJFOQTRK-UHFFFAOYSA-N
1-[4-[2-(Cyclopropylmethoxy)ethyl]phenoxy]-3-isopropylamino-2-propanol hydrochloride
107952
CHEBI:643228

Selective β1 antagonist

Selective β1-adrenoceptor antagonist.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 50mg
SMILES CC(C)NCC(COC1=CC=C(C=C1)CCOCC2CC2)O.Cl
IUPAC Name 1-[4-[2-(cyclopropylmethoxy)ethyl]phenoxy]-3-(propan-2-ylamino)propan-2-ol;hydrochloride
Formula Weight 343.89
Chemical Name or Material Betaxolol hydrochloride
Grade >99%

Recommended Storage : Store at Room Temperature