Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Bevacizumab Chimeric Recombinant Rabbit Monoclonal Antibody (Bevacizumab), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Thermo Scientific MA548045

Encompass

Catalog No. PIMA548045


Add to Cart

Description

Description

Specificity: This antibody binds native and reduced human VEGF (isoform 165).

Bevacizumab Chimeric Recombinant Monoclonal antibody specifically detects Bevacizumab Chimeric in Human samples. It is validated for ELISA, Flow Cytometry, Immunoprecipitation, Inhibition Assays, Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Bevacizumab Chimeric
Recombinant Monoclonal
1 mg/mL
PBS with 0.02% ProClin 300
P05067
Rabbit
Affinity chromatography
RUO
351
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
ELISA, Flow Cytometry, Immunoprecipitation, Inhibition Assays, Western Blot
Bevacizumab
Unconjugated
APP
12-IgG1; Avastin; F(ab)-12 IgG1; Fab-12 IgG1; rhuMAb-VEGF
Amyloid-beta peptide.
200 μg
Primary
Human
Liquid
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit