Learn More
Bevco™ Dura Series Cleanroom ISO 4 Polyurethane Seating, Medium Bench Height, Black
Designed for comfort and long-lasting use in critical environments. Bevco™ Dura Controlled Environment Chair is durable and offers total body support with advanced ergonomic features that improve productivity and reduce fatigue.
Manufacturer:  Bevco™ 7300DC
Catalog No. 19-190-033
Add to cart
Description
Description
  • Adjustable height: 17 to 26 in.
  • Polyurethane seat and back are impervious to water, oil and most chemicals
  • Seat texture is puncture resistant, easy-to-clean and provides non-slip surface
  • Easy-to-use pneumatic height adjustment
  • Waterfall seat to aid lower body circulation
  • Fully adjustable contoured back
Specifications
Specifications
No
Mushroom Glide
Nylon
Polyurethane
17 to 26 in.
Black
Chrome
Controlled Environment Chair
Medium Bench Height