Learn More
Bevco™ Everlast Series Cleanroom ISO 4 Polyurethane Seating, Desk Height, Without Footring
Outfit your facility with Bevco's Everlast Cleanroom ISO 4 Certified Polyurethane Seating that combines advanced ergonomic support with industrial durability. Rugged polyurethane resists stains and damage from punctures, grease, water, and chemicals. Various desk height models available.
Manufacturer:  Bevco™ 7051C1BLK
 View more versions of this product
Catalog No. 19-190-121
Add to cart
Description
Description
  • Everlast Cleanroom Series desk height models are specifically designed for outstanding comfort and long-lasting use in controlled environments
    • These durable chairs offer total body support with advanced ergonomic features that improve productivity and reduce fatigue
    • These Cleanroom models are designed to meet the particulate goals of ISO 14644-1 and are ISO 4 certified by an independent testing laboratory
      • Contoured seat and back have ventilated ribs for lasting comfort and support
      • Durable polyurethane seat and back are impervious to water, oil and most chemicals
      • Easy-to-use pneumatic seat height adjustment with lifetime cylinder guarantee
      • A sturdy 5-legged base ensures stability
      • Adjustable armrests with polyurethane arm pads (available on tilt models only)
      • Available in different colors (black, grey or blue)
      • Dual-wheel hard floors casters are available (add one inch to seat height range)
      • 12 year warranty
      • Supports up to 300 lbs.
      • Meets ANSI/BIFMA standards
      • GreenGuard Gold certified
Specifications
Specifications
Seat Height, Tilt Seat
Mushroom Glide
Polished Aluminum
Polyurethane
14.5 to 19.5 in.
Black
No
Controlled Environment Chair
Desk Height