Learn More
Bevco™ Everlast Series Cleanroom ISO 4 Polyurethane Seating, Desk Height, Without Footring
Outfit your facility with Bevco's Everlast Cleanroom ISO 4 Certified Polyurethane Seating that combines advanced ergonomic support with industrial durability. Rugged polyurethane resists stains and damage from punctures, grease, water, and chemicals. Various desk height models available.
$327.00 - $350.00
Specifications
Axis Type 360° swivel
Certifications ANSI BIFMA, LEED-CI 4.5, Class 10
For Use With (Application) Ideal for comfort and long lasting use in the toughest workplace environment
Lift Type Pneumatic
Seating Height Desk
View More Specs
Products 6
Catalog Number Mfr. No. Color Frame Material Tilt Adjustment (Back) Price Quantity & Availability  
Catalog Number Mfr. No. Color Frame Material Tilt Adjustment (Back) Price Quantity & Availability  
19-130-2860
View Documents
Bevco™
7001C1BLK
Black Reinforced Plastic Articulating
Each for $327.00
Add to cart
19-130-2858
View Documents
Bevco™
7000C1GRY
Gray Reinforced Plastic Manual
Each for $343.00
Add to cart
19-130-2861
View Documents
Bevco™
7001C1BLU
Blue Reinforced Plastic Articulating
Each for $333.00
Add to cart
19-130-2864
View Documents
Bevco™
7001C1GRY
Gray Reinforced Plastic Articulating
Each for $333.00
Add to cart
19-130-2854
View Documents
Bevco™
7000C1BLK
Black Reinforced Plastic Manual
Each for $348.00
Add to cart
19-058-103
View Documents
Bevco™
7000C1BLU
Blue Reinforced Plastic Manual
Each for $350.00
Add to cart
Description
Description
  • Everlast Cleanroom Series desk height models are specifically designed for outstanding comfort and long-lasting use in controlled environments
    • These durable chairs offer total body support with advanced ergonomic features that improve productivity and reduce fatigue
    • These Cleanroom models are designed to meet the particulate goals of ISO 14644-1 and are ISO 4 certified by an independent testing laboratory
      • Contoured seat and back have ventilated ribs for lasting comfort and support
      • Durable polyurethane seat and back are impervious to water, oil and most chemicals
      • Easy-to-use pneumatic seat height adjustment with lifetime cylinder guarantee
      • A sturdy 5-legged base ensures stability
      • Adjustable armrests with polyurethane arm pads (available on tilt models only)
      • Available in different colors (black, grey or blue)
      • Dual-wheel hard floors casters are available (add one inch to seat height range)
      • 12 year warranty
      • Supports up to 300 lbs.
      • Meets ANSI/BIFMA standards
      • GreenGuard Gold certified
Specifications
Specifications
360° swivel
Ideal for comfort and long lasting use in the toughest workplace environment
Desk
Polyurethane
Height
ANSI BIFMA, LEED-CI 4.5, Class 10
Pneumatic
Yes
No