Learn More
BEX1/BEX2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSB00008W
Catalog No. PIOSB00008W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BEX1/BEX2
Polyclonal
Bex2
Bex1, Bex2, Brain expressed gene 1, BEX1, EG2RVC, BEX2, Brain-expressed X-linked protein 2 homolog
Rabbit
100 μL
Primary
Mouse, Rat
Antibody
IgG
Immunohistochemistry, Western Blot
Unconjugated
Q9R224, Q9WTZ8, Q3MKQ1, Q3MKQ2
BEX1, BEX2
A synthetic peptide from n-terminal region of mouse Brain-expressed X-linked protein 1 and 2
RUO
12069, 19716, 363498, 501625
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions
Product Suggestions