Learn More
BEX6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier:  Thermo Scientific OSB00012W
Catalog No. PIOSB00012W
Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reconstitute with 100 μL of distilled water.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
BEX6
Polyclonal
whole serum with no preservative
Q3TZW7
BEX6
A synthetic peptide from c-terminal region of mouse Brain-expressed X-linked protein 6
RUO
328660
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Immunohistochemistry, Western Blot
Unconjugated
Bex6
Brain expressed gene 6, BEX6
Rabbit
100 μL
Primary
Mouse
Antibody
IgG
Recommendations
Recommendations