Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Bexarotene

Potent and selective RXR agonist

$72.70 - $307.00

Specifications

Chemical Name or Material 4-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic acid
Molecular Formula C24H28O2
CAS 153559-49-0
Product Type Target: Retinoid X Receptor Agonists
Formula Weight 348.48
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
58-191-0 View Documents Tocris Bioscience™
5819/10
Each for $72.70
Add to cart
 
58-195-0 View Documents Tocris Bioscience™
5819/50
50mg Each for $307.00
Add to cart
 
Description

Description

Potent and selective RXR agonist (EC50 values are 24, 25 and 33 nM for RXRβ, RXRγ and RXRα, respectively). Exhibits >300-fold selectivity for RXR over RAR receptors. Inhibits tumor cell invasion and migration in vitro, and inhibits metastasis and angiogenesis in mouse models. Also promotes brown adipogenic differentiation and induces brown adipogenic reprogramming from myoblasts.
Specifications

Specifications

4-[1-(5,6,7,8-Tetrahydro-3,5,5,8,8-pentamethyl-2-naphthalenyl)ethenyl]benzoic acid
153559-49-0
348.48
C24H28O2
Target: Retinoid X Receptor Agonists
Safety and Handling

Safety and Handling

Recommended Storage : Store at 4°C

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit