Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHA, FCC, 98.5%, Spectrum™

Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
25013-16-5
C11H16O2
180.25
MRBKEAMVRSLQPH-UHFFFAOYSA-N
2-tert-butyl-4-methoxyphenol
COC1=CC=C(O)C(=C1)C(C)(C)C
25013-16-5
C11H16O2
180.25
MRBKEAMVRSLQPH-UHFFFAOYSA-N
2-tert-butyl-4-methoxyphenol
COC1=CC=C(O)C(=C1)C(C)(C)C

BH104, 25013-16-5, C11H16O2

Spectrum™ BHA, FCC is an antioxidant and preservative used in food, food packaging and animal feed. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Ignition Residue 0.05%
Packaging Amber Glass Bottle
Quantity 125 g
CAS Max % 100%
Percent Purity 98.5%