Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ BHE23 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA568877
Description
This target displays homology in the following species: Mouse: 100%; Rat: 93%.
This gene encodes a member of the basic helix-loop-helix transcription factor family. Members of this family contain two highly conserved and functionally distinct domains: the basic domain targets sequence-specific DNA binding, while the helix-loop-helix domain facilitates protein interaction. Studies of a related gene in mouse suggest that the encoded protein may function as a transcriptional repressor in the pancreas and brain, and that it is required for normal retinal function.
Specifications
BHE23 | |
Polyclonal | |
Unconjugated | |
Bhlhe23 | |
A930001L02Rik; bA305P22.3; basic helix-loop-helix domain containing, class B, 4; basic helix-loop-helix domain containing, class B4; basic helix-loop-helix family member e23; basic helix-loop-helix family, member e23; BETA4; beta-cell E-box transactivating factor 4; BHLHB4; BHLHE23; Class B basic helix-loop-helix protein 4; class E basic helix-loop-helix protein 23; H20; RGD1559760 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
140489 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8BGW3 | |
Bhlhe23 | |
synthetic peptide directed towards the n terminal region of mouse BHE23. | |
100 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction