Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHLHA9 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549086

Catalog No. PA549086

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with mouse based on sequence homology.

Putative transcription factor, which may play a role in limb development.
Specifications

Specifications

BHLHA9
Polyclonal
PBS with 0.09% sodium azide
Q7RTU4
Class A basic helix-loop-helix protein 9, bHLHa9, Class F basic helix-loop-helix factor 42, bHLHf42, BHLHA9, BHLHF42
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
727857
Western Blot
Unconjugated
BHLHA9
Liquid
BHLHA9
KLH conjugated synthetic peptide between 57-83 amino acids from the Central region of human BHLHA9.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit