Learn More
BHLHE41 Mouse anti-Human, Clone: OTI11B5, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF806281
Catalog No. 50-166-7773
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.
Specifications
Specifications
BHLHE41
Monoclonal
Unconjugated
BHLHE41
Lyophilized
BHLHE41
Human recombit protein fragment corresponding to amino acids 1-297 of human BHLHE41 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI11B5
PBS with 8% trehalose and no preservative; pH 7.3
Q9C0J9
BHLHB3, DEC2, SHARP1, hDEC2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
79365