Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BHMT Mouse anti-Human, DyLight 680, Clone: 3D6, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP104264FR
Description
BHMT Monoclonal specifically detects BHMT in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.Specifications
BHMT | |
Monoclonal | |
DyLight 680 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
BHMT | |
Recombinant human BHMT (1-406aa) purified from E. coli | |
0.1 mL | |
Primary | |
NP_001704 | |
Store at 4C in the dark. | |
IgG2a κ |
Western Blot, ELISA | |
3D6 | |
Western Blot, ELISA | |
betaine homocysteine methyltransferase, betaine-homocysteine methyltransferase, betaine--homocysteine S-methyltransferase, betaine--homocysteine S-methyltransferase 1, BHMT1, EC 2.1.1, EC 2.1.1.5 | |
Mouse | |
Protein G purified | |
RUO | |
635 | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title