Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHMT Mouse anti-Human, Janelia Fluor™ 646, Clone: OTI3E11, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270250JF646

Catalog No. N270250J646


Add to cart

Description

Description

BHMT Monoclonal antibody specifically detects BHMT in Human samples. It is validated for Immunohistochemistry.
Specifications

Specifications

BHMT
OTI3E11
Immunohistochemistry
Affinity Purified, Conjugated
BHMT
Full length recombinant protein of human BHMT (NP_001704) produced in HEK293T cells.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry
Janelia Fluor 646
50mM Sodium Borate with 0.05% Sodium Azide
betaine homocysteine methyltransferase, betaine-homocysteine methyltransferase, betaine--homocysteine S-methyltransferase, betaine--homocysteine S-methyltransferase 1, BHMT1, EC 2.1.1, EC 2.1.1.5
Mouse
IgG2a
0.1 mL
Primary
635.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit