Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ BHPI

Click to view available options
Quantity:
10 mg
50 mg
Description
BHPI is an estrogen receptor (ER) α inhibitor. Inhibits proliferation of drug-resistant ERα-positive breast and ovarian cancer cells. Induces sustained activation of the unfolded protein response. Active in vivo.
Specifications
Specifications
Chemical Name or Material | 1,3-Dihydro-3,3-bis(4-hydroxyphenyl)-7-methyl-2H-indol-2-one |
CAS | 56632-39-4 |
Assay Percent Range | >98% |
Quantity | 10 mg |
Molecular Formula | C21H17NO3 |
Formula Weight | Observed MW: 331.36 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction