Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BHQ, Tocris Bioscience™

Click to view available options
Quantity:
100mg
This item is not returnable. View return policy
88-58-4
C14H22O2
222.33
MFCD00008825
JZODKRWQWUWGCD-UHFFFAOYSA-N
2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
2374
CHEBI:41094
2,5-di-tert-butylbenzene-1,4-diol
CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C
This item is not returnable. View return policy
88-58-4
C14H22O2
222.33
MFCD00008825
JZODKRWQWUWGCD-UHFFFAOYSA-N
2,5-di-tert-butylhydroquinone, 2,5-di-tert-butylbenzene-1,4-diol, dibug, dtbhq, dybug, santovar o, di-t-butylhydroquinone, 2,5-di-tert-butylquinol, nonflex alba, 2,5-di-t-butylhydroquinone
2374
CHEBI:41094
2,5-di-tert-butylbenzene-1,4-diol
CC(C)(C)C1=CC(O)=C(C=C1O)C(C)(C)C

Inhibitor of SERCA ATPase

A selective inhibitor of endoplasmic reticulum Ca2+-ATPase.
Quantity 100mg
Formula Weight 222.33
Percent Purity >98%
Chemical Name or Material BHQ

Recommended Storage : Store at Room Temperature