Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Bialaphos sodium, 85%

Click to view available options
Quantity:
100mg
500mg
71048-99-2
C11H21N3NaO6P
345.268
MFCD01674432
RTWIRLHWLMNVCC-WQYNNSOESA-M
118988468
sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate
CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+]
71048-99-2
C11H21N3NaO6P
345.268
MFCD01674432
RTWIRLHWLMNVCC-WQYNNSOESA-M
118988468
sodium;[(3S)-3-amino-4-[[(2S)-1-[[(1S)-1-carboxyethyl]amino]-1-oxopropan-2-yl]amino]-4-oxobutyl]-methylphosphinate
CC(C(=O)NC(C)C(=O)O)NC(=O)C(CCP(=O)(C)[O-])N.[Na+]

Has successfully been used in major cereal species transformation

Applications
Bialaphos Sodium Salt is a tripeptide antibiotic that is employed in the transformation experiments of many species of plants that make use of the bar gene for selection. It is more effective than glufosinate in corn. In wheat, bialaphos was the most reliable selection criteria used to identify bar transformed plants.

Solubility
Soluble in water

Notes
Research Use only: Not intended for animal or human diagnostic or therapeutic use.
UN Number UN2811
Solubility Information Soluble in water
Formula Weight 345.26
Physical Form Powder
Percent Purity 85%
Boiling Point 793°C
Color Orange
Melting Point 160°C
Quantity 500mg
Chemical Name or Material Bialaphos sodium

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : AY6827000

storageNote1 : 2° to 8°C