Learn More
Tocris Bioscience™ BIBO 3304 trifluoroacetate Available on GSA/VA Contract for Federal Government customers only.
Highly selective NPY Y1 receptor antagonist
$328.50 - $1398.00
Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
24-121-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
2412/10
Each for $328.50
Add to cart
24-125-0 Available on GSA/VA Contract for Federal Government customers only. View Documents Tocris Bioscience™
2412/50
Each for $1,398.00
Add to cart
Description
Description
High affinity NPY Y1 receptor antagonist (IC50 values are 0.38 and 0.72 nM at human and rat receptors respectively) that displays > 2600-fold selectivity over Y2, Y4 and Y5 receptors. Inhibits NPY- and fasting-induced feeding in vivo following central administration. Also antagonizes anxiolytic-like effects of NPY.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.