Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIBP 3226 trifluoroacetate, Tocris Bioscience™

Click to view available options
Quantity:
1 mg
10 mg
This item is not returnable. View return policy
1068148-47-9
C29H32F3N5O5
587.6
MTSZIDSCWZHKOD-GNAFDRTKSA-N
bibp 3226 trifluoroacetate, n-1r-4-aminoiminomethyl amino-1-4-hydroxyphenyl methyl amino carbonyl butyl-alpha-phenylbenzeneacetamide trifluoroacetate
56972180
(2R)-5-(diaminomethylideneamino)-2-[(2,2-diphenylacetyl)amino]-N-[(4-hydroxyphenyl)methyl]pentanamide;2,2,2-trifluoroacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)NC(CCCN=C(N)N)C(=O)NCC3=CC=C(C=C3)O.C(=O)(C(F)(F)F)O
This item is not returnable. View return policy
1068148-47-9
C29H32F3N5O5
587.6
MTSZIDSCWZHKOD-GNAFDRTKSA-N
bibp 3226 trifluoroacetate, n-1r-4-aminoiminomethyl amino-1-4-hydroxyphenyl methyl amino carbonyl butyl-alpha-phenylbenzeneacetamide trifluoroacetate
56972180
(2R)-5-(diaminomethylideneamino)-2-[(2,2-diphenylacetyl)amino]-N-[(4-hydroxyphenyl)methyl]pentanamide;2,2,2-trifluoroacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)NC(CCCN=C(N)N)C(=O)NCC3=CC=C(C=C3)O.C(=O)(C(F)(F)F)O

Mixed NPY Y1 and NPFF receptor antagonist

Mixed non-peptide neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist (Ki values are 1.1, 79, 108, > 1000, > 1000 and >1000 for rNPY Y1, hNPFF2, rNPFF, rNPY Y2, rNPY Y4 and rNPY Y5 respectively). Produces an anxiogenic-like effect in rats following i.c.v. administration.
Linear Formula C27H31N5O3.xCF3CO2H
Quantity 1 mg
Formula Weight 473.57
Percent Purity >98%
Chemical Name or Material N-[(1 R)]-4-[(Aminoiminomethyl)amino-1-[[[(4-hydroxyphenyl)methyl]amino]carbonyl]butyl-α-phenylbenzeneacetamide trifluoroacetate

Recommended Storage : Store at -20°C