Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

BIBP 3226 trifluoroacetate, Tocris Bioscience™ Available on GSA/VA Contract for Federal Government customers only.

Click to view available options
Quantity:
1mg
10mg
This item is not returnable. View return policy
1068148-47-9
C29H32F3N5O5
587.6
MTSZIDSCWZHKOD-GNAFDRTKSA-N
bibp 3226 trifluoroacetate,n-1r-4-aminoiminomethyl amino-1-4-hydroxyphenyl methyl amino carbonyl butyl-alpha-phenylbenzeneacetamide trifluoroacetate
56972180
(2R)-5-(diaminomethylideneamino)-2-[(2,2-diphenylacetyl)amino]-N-[(4-hydroxyphenyl)methyl]pentanamide;2,2,2-trifluoroacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)NC(CCCN=C(N)N)C(=O)NCC3=CC=C(C=C3)O.C(=O)(C(F)(F)F)O
This item is not returnable. View return policy
1068148-47-9
C29H32F3N5O5
587.6
MTSZIDSCWZHKOD-GNAFDRTKSA-N
bibp 3226 trifluoroacetate,n-1r-4-aminoiminomethyl amino-1-4-hydroxyphenyl methyl amino carbonyl butyl-alpha-phenylbenzeneacetamide trifluoroacetate
56972180
(2R)-5-(diaminomethylideneamino)-2-[(2,2-diphenylacetyl)amino]-N-[(4-hydroxyphenyl)methyl]pentanamide;2,2,2-trifluoroacetic acid
C1=CC=C(C=C1)C(C2=CC=CC=C2)C(=O)NC(CCCN=C(N)N)C(=O)NCC3=CC=C(C=C3)O.C(=O)(C(F)(F)F)O

Mixed NPY Y1 and NPFF receptor antagonist

Mixed non-peptide neuropeptide Y Y 1 (NPY Y 1 ) and neuropeptide FF (NPFF) receptor antagonist (K i values are 1.1, 79, 108, > 1000, > 1000 and >1000 for rNPY Y 1 hNPFF 2 rNPFF, rNPY Y 2 rNPY Y 4 and rNPY Y 5 respectively). Produces an anxiogenic-like effect in rats following i.c.v. administration.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Quantity 10mg
Formula Weight 473.57
Percent Purity >98%
Chemical Name or Material BIBP 3226 trifluoroacetate

Recommended Storage : Store at -20°C