Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
BIBP 3226 trifluoroacetate, Tocris Bioscience™
Click to view available options
Quantity:
1 mg
10 mg
Description
Mixed non-peptide neuropeptide Y Y1 (NPY Y1) and neuropeptide FF (NPFF) receptor antagonist (Ki values are 1.1, 79, 108, > 1000, > 1000 and >1000 for rNPY Y1, hNPFF2, rNPFF, rNPY Y2, rNPY Y4 and rNPY Y5 respectively). Produces an anxiogenic-like effect in rats following i.c.v. administration.
Specifications
Linear Formula | C27H31N5O3.xCF3CO2H |
Quantity | 1 mg |
Formula Weight | 473.57 |
Percent Purity | >98% |
Chemical Name or Material | N-[(1 R)]-4-[(Aminoiminomethyl)amino-1-[[[(4-hydroxyphenyl)methyl]amino]carbonyl]butyl-α-phenylbenzeneacetamide trifluoroacetate |
Safety and Handling
Recommended Storage | Store at -20°C |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction