BIBU 1361 dihydrochloride, Tocris

Selective inhibitor of EGFR-kinase

Manufacturer: R&D Systems 2417/1

Quantity 1mg

 View more versions of this product

Catalog No. 24-171

  Check Availability
Add to Cart

    Chemical Identifiers

    Assay Percent Range >97%
    Molecular Formula C22H27ClFN7·2HCl
    Formula Weight 516.87
    Synonym N-(3-Chloro-4-fluorophenyl)-6-[4-[(diethylamino)methyl]-1-piperidinyl]-pyrimido[5, 4-d]pyrimidin-4-amine dihydrochloride
    Chemical Name or Material BIBU 1361 dihydrochloride

    Specifications

    Formula Weight 516.87
    Quantity 1mg
    Assay Percent Range >97%
    Molecular Formula C22H27ClFN7·2HCl
    CAS 793726-84-8
    Synonym N-(3-Chloro-4-fluorophenyl)-6-[4-[(diethylamino)methyl]-1-piperidinyl]-pyrimido[5, 4-d]pyrimidin-4-amine dihydrochloride
    Recommended Storage Desiccate at +4°C
    Chemical Name or Material BIBU 1361 dihydrochloride

    Desiccate at +4°C

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit